2-Mercapto-1-methylimidazole
Catalog # EHERC15020200
Supplier: EHRENSTORFER
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:0,25 g
- Seq:3029
Specifications
About this item
:
:114.17 g/mol
Merck Index:13,06000
Boiling Pt:155 °C (1013 hPa)
Density:1.36 g/cm³ (20 °C)
EINECS:200-482-4
CAS Number:60-56-0
Formula:C₄H₆N₂S
MDL Number:MFCD00179321