Order Entry
Netherlands
ContactUsLinkComponent
3-Nitro-o-xylene
3-Nitro-o-xylene
Catalog # EHERC12727900
Supplier:  EHRENSTORFER
CAS Number:  
3-Nitro-o-xylene
Catalog # EHERC12727900
Supplier:  EHRENSTORFER
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    0,25 g
  • Seq:
    3029

Specifications

About this item

 :
 :151.16 g/mol
Boiling Pt:245 °C (1013 hPa)
Density:1.1402 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:201-474-3
UN:1665
ADR:6.1,II
CAS Number:83-41-0
Formula:C₈H₉NO₂
MDL Number:MFCD00007162