
1-(4-Chlorophenyl)-2-thiourea
Supplier: Thermo Scientific
Certificates
About this item
CAS Number:3696-23-9
:
Formula:(C₆H₄Cl)NC(S)NH₂
MDL Number:MFCD00022168
Boiling Pt:304 °C (1013 hPa)
Melting Pt:201…201.5 °C
EINECS:223-022-4
UN:2811
ADR:6.1,II
:186.66 g/mol
:GHS06
Frequently Bought Together