1,3-Dimethyl-2-imidazolidinone
Supplier: Thermo Scientific
Certificates
About this item
CAS Number:80-73-9
:
Formula:C₅H₁₀N₂O
MDL Number:MFCD00003188
:114.15 g/mol
Merck Index:13,03275
Boiling Pt:225.5 °C (1013 hPa)
Melting Pt:8 °C
Density:1.04 g/cm³ (20 °C)
Flash Pt:93 °C
EINECS:201-304-8
UN:2810
ADR:6.1,III
Frequently Bought Together