
1-(3-Chlorophenyl)-2-thiourea
Supplier: Thermo Scientific
Certificates
About this item
CAS Number:4947-89-1
:
Formula:(C₆H₄Cl)NC(S)NH₂
MDL Number:MFCD00022165
Boiling Pt:304 °C (1013 hPa)
Melting Pt:139…140.5 °C
UN:2811
ADR:6.1,II
:186.66 g/mol
:GHS06
Frequently Bought Together