
1-(2-Chlorophenyl)-2-thiourea
Supplier: CHEMPUR
Certificates
About this item
CAS Number:5344-82-1
:
Formula:(C₆H₄Cl)NC(S)NH₂
MDL Number:MFCD00004934
:186.66 g/mol
Boiling Pt:304 °C (1013 hPa)
Melting Pt:147 °C
EINECS:226-291-6
UN:2811
ADR:6.1,I
:GHS06
Frequently Bought Together