
(Benzyl)triphenylphosphonium chloride
Supplier: CHEMPUR
Certificates
About this item
CAS Number:1100-88-5
:
Formula:C₂₅H₂₂ClP
MDL Number:MFCD00011913
:388.88 g/mol
Melting Pt:345…347 °C
Density:1.18 g/cm³ (20 °C)
EINECS:214-154-3
UN:3464
ADR:6.1,II
:GHS06
Frequently Bought Together