2'-Hydroxy-4'-methoxyacetophenone
Supplier: Thermo Scientific
Certificates
About this item
CAS Number:552-41-0
:
Formula:C₉H₁₀O₃
MDL Number:MFCD00008730
:166.18 g/mol
Boiling Pt:154 °C (20 mmHg)
Melting Pt:51 °C
Density:1,1604 g/cm³ (40 °C)
EINECS:209-012-2
Frequently Bought Together