Bovine thrombin (from plasma)
Supplier: Merck Millipore (Calbiochem)
Certificates
About this item
Specifications
- CAS No.:9002-04-4
- Source:Plasma
- Species:Bovine
- Form:Lyophilised
- Enzyme name:Thrombin
- Purity:Complete activation from homogeneous prothrombin by SDS-PAGE
- Formulation:Lyophilised from 200 mM NaCl, 50 mM sodium citrate, 1 ml 0,1% PEG, pH 6,5
Specifications
Frequently Bought Together