Order Entry
Netherlands
Orders LinkContactUsLinkComponent
Sodium cholate, Millipore®

Sodium cholate, Millipore®

Glass bottle for solids, 50 g
Catalog # 229101-50
Sodium cholate, Millipore®
Catalog # 229101-50
CAS Number:  361-09-1
Sodium cholate, Millipore®

Glass bottle for solids, 50 g

CAS Number:  361-09-1
Price
170,00 €
Log in for availability
1 * 50 g
Product Details & Documents
Synonyms:  3α,7α,12α-Trihydroxy-5β-cholanic acid sodium salt, 3α,7α,12α-Trihydroxycholan-24-oic acid sodium salt, 3α,7α,12α-Trihydroxycholanic acid sodium salt, Sodium 3α,7α,12α-trihydroxy-5β-cholanoate, (R)-4-((3R,5S,7R,8R,9S,10S,12S,13R,14S,17R)-3,7,12-Trihydroxy-10,13-dimethylhexadecahydro-1H-cyclopenta[a]phenanthren-17-yl)pentanoic acid sodium salt, 3α,7α,12α-Trihydroxy-5β-cholan-24-oic acid sodium salt
  • CAS Number:361-09-1
  • Formula:C₂₄H₃₉NaO₅
  • MDL Number:MFCD00150749
  • Molecular Weight:430.56 g/mol
  • Melting Pt:198 °C
  • Storage Temperature:Ambient
  • EINECS:206-643-5
Specifications

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    50 g

Specifications