


2-Chloro-5-(trifluoromethyl)pyridine ≥97%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:52334-81-3
:
Formula:C₆H₃ClF₃N
MDL Number:MFCD00042225
:181.54 g/mol
Boiling Pt:152…153 °C
Melting Pt:29…33 °C
Density:1.417
Flash Pt:62 °C (143 °F)
Storage Temperature:Ambient
EINECS:257-856-5
UN:2811
ADR:6.1,III
:GHS06
:GHS08
:GHS07
Related Information
Frequently Bought Together