

3-(Trifluoromethyl)benzyl mercaptan tech. 90%, Technical Grade
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:25697-55-6
:
Formula:C₈H₇F₃S
MDL Number:MFCD00042435
:192.2 g/mol
Boiling Pt:75…77 °C (5 mmHg)
UN:2810
ADR:6.1,III
:GHS06
:GHS07
Storage Temperature:Ambient
Related Information
Frequently Bought Together