Order Entry
Netherlands
ContactUsLinkComponent
5-(2,3-Dichlorophenyl)-1H-tetrazole ≥97%
Irritant
5-(2,3-Dichlorophenyl)-1H-tetrazole ≥97%
Catalog # B20921.06
CAS Number:  
Irritant
5-(2,3-Dichlorophenyl)-1H-tetrazole ≥97%
Catalog # B20921.06
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    5 g

Specifications

About this item

 :
 :215.04 g/mol
Melting Pt:180…182 °C
 :GHS07
Storage Temperature:Ambient
CAS Number:175205-12-6
Formula:C₇H₄Cl₂N₄
MDL Number:MFCD00068096