
1-Naphtholbenzeine
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:145-50-6
:6948-88-5
Formula:C₂₇H₁₈O₂
MDL Number:MFCD00078492
:374.44 g/mol
Melting Pt:∼. 225 °C (dec.)
EINECS:205-656-3
:GHS07
Storage Temperature:Ambient
Related Information
Frequently Bought Together