
Trimethylsulphoxonium iodide ≥98%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:1774-47-6
:
Formula:C₃H₉IOS
MDL Number:MFCD00011899
:220.07 g/mol
Melting Pt:∼ 170 °C (dec.)
EINECS:217-204-2
:GHS07
Storage Temperature:Ambient
Related Information
Frequently Bought Together