Gadolinium, sputtering target, Ø 50.8 mm, Thickness: 3.18 mm
Catalog # 45737.KS
Supplier: Thermo Fisher Scientific
CAS Number:
Specifications
- Pk:1
- Seq:0001
Specifications
About this item
:
:157.25 g/mol
Storage Temperature:Ambient
EINECS:231-162-2
CAS Number:7440-54-2
Formula:Gd
MDL Number:MFCD00011022