N-Ethylbenzylamine 95%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:14321-27-8
:
Formula:C₆H₅CH₂NHCH₂CH₃
MDL Number:MFCD00009031
:135.21 g/mol
Boiling Pt:191…194 °C (760 mmHg)
Density:0.9 g/cm³
Flash Pt:66 °C
Storage Temperature:Ambient
EINECS:238-265-1
UN:2735
ADR:8,II
:GHS05
Specification Test Results
- Appearance:Clear colorless to light yellow Liquid
- Infrared spectrum:Conforms
- Refractive index:1.5100 to 1.5130 (20°C, 589 nm)
- Titration with HClO4:≥94 %
Specification Test Results
Frequently Bought Together