

Trichloromethyl chloroformate 99%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:503-38-8
:
Formula:C₂Cl₄O₂
MDL Number:MFCD00015553
:197.83 g/mol
Merck Index:13,03368
Boiling Pt:128 °C (760 mmHg)
Melting Pt:–57 °C
Density:1.64 g/cm³
Storage Temperature:Refrigerator
EINECS:207-965-9
UN:3390
ADR:6.1,
:GHS06
:GHS05
Specification Test Results
- Appearance:Clear colorless Liquid
- Infrared spectrum:Conforms
- Refractive index:1.4560 to 1.4590 (20°C, 589 nm)
- GC:≥98.5 %
Specification Test Results
Frequently Bought Together