

(Chloromethyl)trimethylsilane 98+%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:2344-80-1
:
Formula:ClH₂CSi(CH₃)₃
MDL Number:MFCD00000878
:122.67 g/mol
Boiling Pt:98 °C (1013 hPa)
Density:0.879 g/cm³
Flash Pt:–2 °C
EINECS:219-058-5
UN:1993
:GHS07
:GHS02
Specification Test Results
- Appearance:Clear colorless Liquid
- Infrared spectrum:Conforms
- Refractive index:1.4165 to 1.4185 (20°C, 589 nm)
- GC:≥98.0 %
Specification Test Results
Frequently Bought Together