Order Entry
Netherlands
ContactUsLinkComponent
Tricine 98% for biochemistry
Irritant
Tricine 98% for biochemistry
Certificates
undefined
Irritant
Tricine 98% for biochemistry

Specification Test Results

  • Appearance:
    White Powder
  • Infrared spectrum:
    Conforms
  • Loss on drying:
    ≤1 %
  • UV:
    (0.1 M in water)
  • pH:
    4.0 to 6.0 (1 % in water at 20°C)
  • Titration with HClO4:
    ≥97.5 % (On dried basis)
  • Heavy metals (as Pb):
    ≤5 ppm

Specification Test Results

Related Information

Frequently Bought Together