

Trimethylsulphoxonium iodide 98%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:1774-47-6
:
Formula:C₃H₉IOS
MDL Number:MFCD00011899
:220.07 g/mol
Melting Pt:175 °C (dec.)
EINECS:217-204-2
:GHS07
:GHS09
Specification Test Results
- Appearance:White to light yellow Crystalline powder
- Infrared spectrum:Conforms
- Titration Argentometric:≥97.5 %
- Specific optical rotation:-142° to -146° (20°C, 589 nm) (c=1, ethanol)
Specification Test Results
Frequently Bought Together