

2-Mercapto-1-methylimidazole 98%
:60-56-0
:
:C₄H₆N₂S
:MFCD00179321
:114.17 g/mol
:13,06000
:280 °C (760 mmHg)
:140…145 °C
:1,36 g/cm³ (20 °C)
:>100 °C
:Ambient
:200-482-4
:3335
:9,III
:GHS07
:GHS08
- Appearance:White to cream to yellow-brown Crystalline powder
- Infrared spectrum:Conforms
- Melting point:140 to 145 °C
- Loss on drying:≤1 % (105 °C, 2 hrs)
- Titration with NaOH:≥97.5 % (On dried substance)
Frequently Bought Together