Order Entry
Netherlands
ContactUsLinkComponent
2,6-Dichlorobiphenyl (PCB No. 10)
2,6-Dichlorobiphenyl (PCB No. 10)
Catalog # ACCUC-010N
Supplier:  AccuStandard
CAS Number:  
2,6-Dichlorobiphenyl (PCB No. 10)
Catalog # ACCUC-010N
Supplier:  AccuStandard
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    25 mg
  • Seq:
    2315

Specifications

About this item

 :
ADR:9,II
 :223.1 g/mol
UN:3432
CAS Number:33146-45-1
Formula:C₁₂H₈Cl₂