
Methyl 4-bromo-2-fluoro-6-(trifluoromethyl)benzoate 95%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:2091662-46-1
:
Formula:C₉H₅BrF₄O₂
MDL Number:MFCD33404168
:301.03 g/mol
:GHS07
Frequently Bought Together
Ethanol, GPR RECTAPUR®
Supplier: VWR Chemicals
2-Propanol ≥99.0%, GPR RECTAPUR®
Supplier: VWR Chemicals
Water with 0,1% (v/v) trifluoroacetic acid, HiPerSolv CHROMANORM® for LC-MS
Supplier: VWR Chemicals
Ethanol absolute ≥99.8%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
Supplier: VWR Chemicals
Ethanol absolute ≥99.8%, HiPerSolv CHROMANORM®, gradient grade for HPLC
Supplier: VWR Chemicals
1,4-Dioxane ≥99.5% stabilised, AnalaR NORMAPUR® analytical reagent
Supplier: VWR Chemicals
Toluene ≥99.90%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
Supplier: VWR Chemicals
Methanol ≥99.8%, HiPerSolv CHROMANORM® Reag. Ph. Eur., gradient grade for HPLC
Supplier: VWR Chemicals
Water, TECHNICAL, demineralised
Supplier: VWR Chemicals
1-Butanol ≥98.5%, GPR RECTAPUR®
Supplier: VWR Chemicals
Dichloromethane, dehydrated (max. 0.01% H₂O) ≥99% stabilised, GPR RECTAPUR® for synthesis
Supplier: VWR Chemicals
Glycerine (glycerol) USP, Ph. Eur., BP
Supplier: VWR Chemicals
2-Propanol ≥98%, TECHNICAL
Supplier: VWR Chemicals
Ethylene glycol ≥99.0% Reag. Ph. Eur.
Supplier: VWR Chemicals
N,N-Dimethylformamide, extra dry over Molecular Sieve 99.8%, AcroSeal®
Supplier: Thermo Fisher Scientific