Order Entry
Netherlands
ContactUsLinkComponent
Methyl 4-bromo-7-fluorobenzo[b]thiophene-2-carboxylate 95%
Irritant
Methyl 4-bromo-7-fluorobenzo[b]thiophene-2-carboxylate 95%
Catalog # APOSPC57446-1G
Supplier:  Apollo Scientific
CAS Number:  
Irritant
Methyl 4-bromo-7-fluorobenzo[b]thiophene-2-carboxylate 95%
Catalog # APOSPC57446-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :289.12 g/mol
 :GHS07
CAS Number:360576-05-2
Formula:C₁₀H₆BrFO₂S
MDL Number:MFCD13180865