Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Bestelling invoeren
Status van de bestelling
Uw account
Winkelwagen
Netherlands
Bestelling invoeren
Status van de bestelling
Uw account
Winkelwagen
ContactUsLinkComponent
Home
Methyl 2-bromo-5-chloro-4-fluorobenzoate
Methyl 2-bromo-5-chloro-4-fluorobenzoate
Methyl 2-bromo-5-chloro-4-fluorobenzoate
Methyl 2-bromo-5-chloro-4-fluorobenzoate
APOSPC49458-1G
:
Apollo Scientific
:
Methyl 2-bromo-5-chloro-4-fluorobenzoate
APOSPC49458-1G
:
Apollo Scientific
:
Pack type:
Bottle
Pk:
1 g
Seq:
3047
:
:
267.48 g/mol
:
GHS07
:
1502835-28-0
:
C₈H₅BrClFO₂
:
MFCD24108550