Supelco® Organic Reference Standard, 4-Methylphenol Solution
Catalog # 40252-U
Supplier: Merck
CAS Number:
Specifications
- Pk:1
- Standards:Certified reference material, 5000 mug/ml in methanol
- CAS No.:106-44-5
- Description:4-Methylphenol solution
- Storage conditions:2...8 °C
- Hazard class description:H225 - H301 + H311 + H331 - H370
- Shipping conditions:STD
- M-Clarity quality level:MQ100
- Synonym(s):4-Methylphenol solution; p-Cresol solution
- Formula:C₇H₈O
Specifications
About this item
- GHS signal: GHS06, GHS08
Caution: P210 - P280 - P301 + P310 + P330 - P302 + P352 + P312 - P304 + P340 + P311