Order Entry
Netherlands
ContactUsLinkComponent
 
Supelco® Organic Reference Standard, o-Cresol 5,000 µg/ml in Methanol, Supelco
  40250-U
 :  Merck
 :  
Supelco® Organic Reference Standard, o-Cresol 5,000 µg/ml in Methanol, Supelco
  40250-U
 :  Merck
 :  

 

  • Pk:
    1
  • CAS No.:
    95-48-7
  • Description:
    o-Cresol 5,000 µg/ml in methanol
  • Formula:
    C₇H₈O