Order Entry
Netherlands
ContactUsLinkComponent
 
Organic reference standard, 1,1-Dichloroethylene 1000 µg/ml in methanol, Supelco®
  SUPL40027
 :  Merck
 :  
Organic reference standard, 1,1-Dichloroethylene 1000 µg/ml in methanol, Supelco®
  SUPL40027
 :  Merck
 :  

 

  • Pk:
    1
  • CAS No.:
    75-35-4
  • Description:
    1,1-Dichloroethylene 1,000 µg/ml in methanol
  • Formula:
    C₂H₂Cl₂
  • Description:
    1,1-Dichloroethylene