

(±)-6-Hepten-3-ol ≥99%
Catalog # B20128.03
Supplier: Thermo Fisher Scientific
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:1 g
Specifications
About this item
CAS Number:19781-77-2
:
Formula:C₇H₁₄O
:114.19 g/mol
Storage Temperature:Ambient
UN:1987
ADR:3,III
:GHS02
:GHS07
MDL Number:MFCD00046683