Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Netherlands
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
FMOC-Glu(TF3)-OH
FMOC-Glu(TF3)-OH for biomolecular labeling
FMOC-Glu(TF3)-OH for biomolecular labeling
FMOC-Glu(TF3)-OH for biomolecular labeling
AATB5016
:
AAT Bioquest
:
FMOC-Glu(TF3)-OH for biomolecular labeling
AATB5016
:
AAT Bioquest
:
Pack type:
Plastic bottle
Pk:
100 mg
Seq:
2440
:
:
5,II
:
GHS07