Bestelling invoeren
Netherlands
ContactUsLinkComponent
D-(+)-Biotin ≥90%
Irritant
D-(+)-Biotin ≥90%
Catalog # AATB3001
Supplier:  AAT BIOQUEST
CAS Number:  
undefined
Irritant
D-(+)-Biotin ≥90%
Catalog # AATB3001
Supplier:  AAT BIOQUEST
CAS Number:  

Specifications

  • Verpakking:
    Plastic bottle
  • VPE:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :244.31 g/mol
ADR:5,II
 :GHS07
CAS Number:58-85-5
Formula:C₁₀H₁₆N₂O₃S
MDL Number:MFCD00005541