Order Entry
Netherlands
ContactUsLinkComponent
Cyanine 5 bissuccinimidyl ester potassium salt ≥90% [same as GE Cy5® bisNHS ester]
Cyanine 5 bissuccinimidyl ester potassium salt ≥90% [same as GE Cy5® bisNHS ester]
Catalog # AATB282
Supplier:  AAT Bioquest
CAS Number:  
undefined
Cyanine 5 bissuccinimidyl ester potassium salt ≥90% [same as GE Cy5® bisNHS ester]
Catalog # AATB282
Supplier:  AAT Bioquest
CAS Number:  

Specifications

  • Pk:
    1 mg
  • Seq:
    2070

Specifications

About this item

 :
 :975.13 g/mol