Acridinium C2 NHS Ester for biomolecular labeling
Supplier: AAT BIOQUEST
New Product
Certificates
About this item
Synonyms: 4-(2-Succinimidyl-oxycarbonylethyl)-phenyl-10-acridinium-9-carboxylate Triflurormethyl sulphonate, Acridinium C2 N-hydroxysuccinimide ester
CAS Number:177332-37-5
:
Formula:C₂₈H₂₃N₂O₆·CF₃O₃S
Molecular Weight:632.56 g/mol
ADR:5 II
:GHS07
Specifications
- Environmentally Preferable:
Specifications
Frequently Bought Together