
Tide Fluor™ 6WS succinimidyl ester, Tide Fluor™ fluorescent probe
Catalog # AATB2294
Supplier: AAT Bioquest
CAS Number:
Specifications
- Pack type:Plastic bottle
- Pk:1 mg
- Seq:2070
Specifications
About this item
:
:1317.7 g/mol
ADR:5,II
:GHS07