0,00 €
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Netherlands
ContactUsLinkComponent
Tide Fluor™ 5WS succinimidyl ester
Tide Fluor™ 5WS succinimidyl ester, Tide Fluor™ fluorescent probe
Tide Fluor™ 5WS succinimidyl ester, Tide Fluor™ fluorescent probe
AATB2281
:
AAT Bioquest
:
Tide Fluor™ 5WS succinimidyl ester, Tide Fluor™ fluorescent probe
AATB2281
:
AAT Bioquest
:
Pack type:
Plastic bottle
Pk:
5 mg
Seq:
2210
:
:
1050.35 g/mol
:
5,II
:
GHS07