Order Entry
Netherlands
ContactUsLinkComponent
Tide Quencher™ 3 succinimidyl ester, Tide Quencher™ fluorescent probe
Irritant
Tide Quencher™ 3 succinimidyl ester, Tide Quencher™ fluorescent probe
Catalog # AATB2230
Supplier:  AAT Bioquest
CAS Number:  
undefined
Irritant
Tide Quencher™ 3 succinimidyl ester, Tide Quencher™ fluorescent probe
Catalog # AATB2230
Supplier:  AAT Bioquest
CAS Number:  

Specifications

  • Pack type:
    Plastic bottle
  • Pk:
    25 mg
  • Seq:
    2315

Specifications

About this item

 :
ADR:5,II
 :550.59 g/mol
 :GHS07