Order Entry
Netherlands
ContactUsLinkComponent
Calcein AM ≥90%
Calcein AM ≥90%
Catalog # AATB22002
Supplier:  AAT BIOQUEST
CAS Number:  
undefined
Calcein AM ≥90%
Catalog # AATB22002
Supplier:  AAT BIOQUEST
CAS Number:  

Specifications

  • Pack type:
    Plastic bottle
  • Pk:
    1 mg
  • Seq:
    2070

Specifications

About this item

 :
 :994.86 g/mol
ADR:5,II
 :GHS07
CAS Number:148504-34-1
Formula:C₄₆H₄₆N₂O₂₃
MDL Number:MFCD05861516