Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Inloggen
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Bestelling invoeren
Status van de bestelling
Uw account
Winkelwagen
Netherlands
Bestelling invoeren
Status van de bestelling
Uw account
Winkelwagen
ContactUsLinkComponent
Home
RH795
RH795 ≥95%
RH795 ≥95%
Irritant
RH795 ≥95%
Catalog # AATB21495
Supplier:
AAT BIOQUEST
CAS Number:
Expand image
Irritant
RH795 ≥95%
Catalog # AATB21495
Supplier:
AAT BIOQUEST
CAS Number:
Specifications
Pack type:
Plastic bottle
Pk:
1 mg
Seq:
2070
Specifications
About this item
Product Details
:
ADR:
5,II
:
585.42 g/mol
:
GHS07
CAS Number:
172807-13-5
Formula:
C₂₆H₃₉Br₂N₃O₂
Product Details