Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Netherlands
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Fura-FF, AM
Fura-FF, AM ≥95%
Fura-FF, AM ≥95%
AATB21027
:
AAT BIOQUEST
:
Fura-FF, AM ≥95%
AATB21027
:
AAT BIOQUEST
:
Pack type:
Plastic bottle
Pk:
500 µG
Seq:
1418
:
:
1023.8 g/mol
:
5,II
:
GHS07
:
348079-12-9
:
C₄₃H₄₃F₂N₃O₂₄