
5-Carboxy-4'-aminomethylfluorescein DNA sequencing terminator building block
Catalog # AATB207
Supplier: AAT Bioquest
CAS Number:
Specifications
- Pack type:Plastic bottle
- Pk:10 mg
- Seq:2275
Specifications
About this item
:
:441.82 g/mol
ADR:5,II
:GHS07