Order Entry
Netherlands
ContactUsLinkComponent
 
Irritant
Tide Quencher™ 2WS succinimidyl ester, Tide Quencher™ fluorescent probe
Catalog # AATB2058
Supplier:  AAT Bioquest
CAS Number:  
undefined
Irritant
Tide Quencher™ 2WS succinimidyl ester, Tide Quencher™ fluorescent probe
Catalog # AATB2058
Supplier:  AAT Bioquest
CAS Number:  

Specifications

  • Pack type:
    Plastic bottle
  • Pk:
    5 mg
  • Seq:
    2210

Specifications

About this item

 :
 :688.86 g/mol
ADR:5,II
 :GHS07