Order Entry
Netherlands
Orders LinkContactUsLinkComponent
 

iFluor® 790, Amine

1 mg
Catalog # AATB1362
Supplier:   AAT Bioquest
iFluor® 790, Amine
Catalog # AATB1362
Supplier:  AAT Bioquest
CAS Number:  
iFluor® 790, Amine
iFluor® 790, Amine

1 mg

CAS Number:  
Price
469,00 €
Log in for availability
1 * 1 mg
Product Details & Documents

In vivo fluorescence imaging uses a sensitive camera to detect fluorescence emission from fluorophores in whole-body living small animals.

  • Bright Fluorescence
  • High Photostability
  • Low Quenching On Proteins
  • Useful For In-Vivo Imaging

 : For research use only.

  • Storage Temperature:Freezer
Specifications

Specifications

  • Pk:
    1 mg

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.