Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Netherlands
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Blue 630 succinimidyl ester
Blue 630 succinimidyl ester, mFluor™ fluorescent probe
Blue 630 succinimidyl ester, mFluor™ fluorescent probe
AATB1164
:
AAT BIOQUEST
:
Blue 630 succinimidyl ester, mFluor™ fluorescent probe
AATB1164
:
AAT BIOQUEST
:
Pack type:
Plastic bottle
Pk:
1 mg
Seq:
2070
:
:
709.86 g/mol
:
5,II
:
GHS07