Order Entry
Netherlands
ContactUsLinkComponent
Violet 510 succinimidyl ester, mFluor™ fluorescent probe
Irritant
Violet 510 succinimidyl ester, mFluor™ fluorescent probe
Catalog # AATB1151
Supplier:  AAT BIOQUEST
CAS Number:  
undefined
Irritant
Violet 510 succinimidyl ester, mFluor™ fluorescent probe
Catalog # AATB1151
Supplier:  AAT BIOQUEST
CAS Number:  

Specifications

  • Pack type:
    Plastic bottle
  • Pk:
    1 mg
  • Seq:
    2070

Specifications

About this item

 :
ADR:5,II
 :770.83 g/mol
 :GHS07