Order Entry
Netherlands
ContactUsLinkComponent
MitoLite™ Orange FX570
  AATB22676
 :  
undefined
MitoLite™ Orange FX570
  AATB22676
 :  
  • Pk:
    500 Tests
  • Description:
    MitoLite™ orange
  • Purity:
    90%
MitoLite™ orange is a cationic dye that selectively accumulates in mitochondria probably vial the mitochondrial membrane potential gradient.
  • Mitochondria-specific dye
  • Cell-permeable
  • Robust
 : For research use only.