Order Entry
Netherlands
ContactUsLinkComponent
Cell Meter™ RX700 Fixable Viability Dye
Cell Meter™ RX700 Fixable Viability Dye
Catalog # AATB22532
Supplier:  AAT BIOQUEST
CAS Number:  
undefined
Cell Meter™ RX700 Fixable Viability Dye
Catalog # AATB22532
Supplier:  AAT BIOQUEST
CAS Number:  
Specifications
  • Pk:
    200 Tests
  • Description:
    Cell Meter™ RX700 fixable cell stain
  • Tests per kit:
    200 Tests
Specifications
About this item
Discrimination and exclusion of dead cells from live cells allows cleaner separation and identification of cell populations.
  • Cell-impermeable
  • Amine- and thiol-reactive
  • High stability
  • Live/dead cell labeling dye
  • Fixable
Caution: For research use only.