Order Entry
Netherlands
ContactUsLinkComponent
Amplite® Choline Quantitation Kit
Amplite® Choline Quantitation Kit
Catalog # AATB40007
Supplier:  AAT Bioquest
CAS Number:  
undefined
Amplite® Choline Quantitation Kit
Catalog # AATB40007
Supplier:  AAT Bioquest
CAS Number:  
Specifications
  • Pk:
    200 Tests
  • Description:
    Amplite™ choline quantitation kit
Specifications
About this item
Choline and its metabolites play an important role in the structural integrity and signaling of cell membranes and cholinergic neurotransmission (acetylcholine synthesis).
  • In-house QC testing
  • Sensitive
  • Suggested protocol available
Caution: For research use only.