Order Entry
Netherlands
ContactUsLinkComponent
Native Human arginine-glycine-aspartic acid (from Human), iFluor® 750
Native Human arginine-glycine-aspartic acid (from Human), iFluor® 750
Catalog # AATB36800
Supplier:  AAT BIOQUEST
CAS Number:  
undefined
Native Human arginine-glycine-aspartic acid (from Human), iFluor® 750
Catalog # AATB36800
Supplier:  AAT BIOQUEST
CAS Number:  

Specifications

  • Pack type:
    Vial
  • Pk:
    1 mg
  • Conjugation:
    iFluor® 750
  • Protein/peptide type:
    Native
  • Species:
    Human
  • Protein synonyms:
    RGD
  • Protein/peptide name:
    arginine-glycine-aspartic acid

Specifications

About this item

Near-infrared fluorescence optical imaging (also called in vivo Imaging) is emerging as a powerful technology for studying diseases at molecular level in small animal models.

  • Nir emission (757/779 nm)
  • Brighter and more stable signal
  • Multiple research applications
  • Suitable for both in vivo and in vitro
Caution: For research use only.