Native Biotin (from Eukaryotic cells), Cal-520®
AATB20605
:
- Pack type:Vial
- Pk:250 µG
- Conjugation:Cal-520®
- Protein/peptide type:Native
- Source:Eukaryotic cells
- Protein synonyms:Vitamin H|B7
- Protein/peptide name:Biotin
Biotin and biocytin bind with high affinity to both avidins and streptavidins.
- High specificity
- Sensitive detection
- Multiple applications
: For research use only.